3,5-Difluoro-4-chlorophenol ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-3,5-difluorophenol

073436-250MG 073436-1G 073436-5G
102530-154EA 400 USD
102530-154 102530-156 102530-158
3,5-Difluoro-4-chlorophenol ≥95%
3,5-Difluoro-4-chlorophenol

Hazard: Irritant

Formula: C₆H₃ClF₂O
MW: 164.54 g/mol
MDL Number: MFCD06658279
CAS Number: 2268-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR