7-Chloro-2-hydroxyquinoline ≥95%

Supplier: Matrix Scientific

073435-250MG 073435-1G 073435-5G
102530-148EA 498.46 USD
102530-148 102530-150 102530-152
7-Chloro-2-hydroxyquinoline ≥95%
7-Chloro-2-hydroxyquinoline

Hazard: Irritant

Formula: C₉H₆ClNO
MW: 179.6030 g/mol
MDL Number: MFCD08460109
CAS Number: 22614-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR