2-Chloro-5-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

073434-1G 073434-5G 073434-10G
102530-142EA 364.62 USD
102530-142 102530-144 102530-146
2-Chloro-5-methoxypyrimidine ≥95%
2-Chloro-5-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.56 g/mol
MDL Number: MFCD08702770
CAS Number: 22536-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR