2-Fluoro-4-pyridone ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Fluoropyridin-4-ol, 2-Fluoro-4-hydroxypyridine
073432-250MG
073432-1G
073432-5G
102530-130EA
498.46
USD
102530-130
102530-132
102530-134
2-Fluoro-4-pyridone ≥95%
2-Fluoro-4-pyridone
Hazard: Irritant
Formula:
C₅H₄FNO MW: 113.09 g/mol |
MDL Number:
MFCD04114159 CAS Number: 22282-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...