Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3,5-dimethyl-2-pyrrolecarboxylate

073430-1G 073430-5G 073430-10G
102530-118EA 72.31 USD
102530-118 102530-120 102530-122
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate ≥95%
Ethyl-3,5-dimethyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00010645
CAS Number: 2199-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR