2,5-Dibromo-4-methylimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Dibromo-4-methyl-1H-imidazole

073429-1G 073429-5G 073429-10G
102530-112EA 533.85 USD
102530-112 102530-114 102530-116
2,5-Dibromo-4-methylimidazole ≥95%
2,5-Dibromo-4-methylimidazole

Hazard: Irritant

Formula: C₄H₄Br₂N₂
MW: 239.9 g/mol
MDL Number: MFCD02179519
CAS Number: 219814-29-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR