5-(Bromomethyl)-2-chloropyridine ≥95%

Supplier: Matrix Scientific

073420-1G 073420-5G
102530-066EA 640 USD
102530-066 102530-068
5-(Bromomethyl)-2-chloropyridine ≥95%
5-(Bromomethyl)-2-chloropyridine

Hazard: Irritant

Formula: C₆H₅BrClN
MW: 206.4700 g/mol
MDL Number: MFCD07368893
CAS Number: 182924-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR