1-Methyl-1H-benzo[d]imidazole-6-carbaldehyde ≥95%
Supplier: Matrix Scientific
073419-250MG
073419-1G
102530-062EA
1350.77
USD
102530-062
102530-064
1-Methyl-1H-benzo[d]imidazole-6-carbaldehyde ≥95%
1-Methyl-1H-benzo[d]imidazole-6-carbaldehyde
Hazard: Irritant
Formula:
C₉H₈N₂O MW: 160.172 g/mol |
MDL Number:
MFCD11977687 CAS Number: 181867-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...