trans-N-Boc-1,4-cyclohexanediamine ≥95%

Supplier: Matrix Scientific

Synonyms: trans-4-(Boc-amino)cyclohexylamine, trans-tert-Butyl (4-aminocyclohexyl)carbamate

073418-1G 073418-5G
102530-058EA 33.85 USD
102530-058 102530-060
trans-N-Boc-1,4-cyclohexanediamine ≥95%
trans-N-Boc-1,4-cyclohexanediamine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.30 g/mol
MDL Number: MFCD03001719
CAS Number: 177906-48-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR