5-Thiazolamine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminothiazole hydrochloride

073417-1G 073417-5G
102530-054EA 783.08 USD
102530-054 102530-056
5-Thiazolamine hydrochloride ≥95%
5-Thiazolamine hydrochloride

Hazard: Irritant

Formula: C3H5ClN2S
MW: 136.6 g/mol
MDL Number: MFCD10687116
CAS Number: 942631-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR