2-Chloropyrimidine-5-carbonitrile ≥95%

Supplier: Matrix Scientific

073414-1G 073414-5G
102530-042EA 113.85 USD
102530-042 102530-044
2-Chloropyrimidine-5-carbonitrile ≥95%
2-Chloropyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₂ClN₃
MW: 139.5400 g/mol
Melting Pt: 140-142°C
MDL Number: MFCD10697145
CAS Number: 1753-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR