6-Methyl-2,4-dihydroxyquinoline ≥95%

Supplier: Matrix Scientific

073413-250MG 073413-1G 073413-5G
102530-034EA 598.46 USD
102530-034 102530-036 102530-038
6-Methyl-2,4-dihydroxyquinoline ≥95%
6-Methyl-2,4-dihydroxyquinoline

Hazard: Irritant

Formula: C₁₀H₉NO₂
MW: 175.1800 g/mol
MDL Number: MFCD00227473
CAS Number: 1677-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR