4,5-Diamino-6-hydroxypyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5,6-Diamino-4-hydroxypyrimidine, 5,6-Diamino-4-pyrimidinol, 5,6-Diamino-4(1H)-pyrimidinone

073412-1G 073412-5G
102530-028EA 36.92 USD
102530-028 102530-030
4,5-Diamino-6-hydroxypyrimidine ≥95%
4,5-Diamino-6-hydroxypyrimidine

Hazard: Irritant

Formula: C₄H₆N₄O
MW: 126.1200 g/mol
Melting Pt: 241 °C
MDL Number: MFCD00006114
CAS Number: 1672-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR