2-Fluoro-4-iodo-3-methylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Fluoro-4-iodo-3-picoline
073409-250MG
073409-1G
073409-5G
102530-012EA
400
USD
102530-012
102530-014
102530-016
2-Fluoro-4-iodo-3-methylpyridine ≥95%
2-Fluoro-4-iodo-3-methylpyridine
Hazard: Irritant
Formula:
C₆H₅FIN MW: 237.02 g/mol |
MDL Number:
MFCD03095289 CAS Number: 153034-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...