4,7-Dibromo-2,1,3-benzothiadiazole ≥95%

Supplier: Matrix Scientific

073407-5G 073407-25G 073407-100G
102530-002EA 41.54 USD
102530-002 102551-834 102551-836
4,7-Dibromo-2,1,3-benzothiadiazole ≥95%
4,7-Dibromo-2,1,3-benzothiadiazole

Hazard: Toxic

Formula: C₆H₂Br₂N₂S
MW: 293.97 g/mol
Melting Pt: 186-190°C
MDL Number: MFCD00658844
CAS Number: 15155-41-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR