N-(2-Chloropyridin-4-yl)acetamide ≥95%

Supplier: Matrix Scientific

073403-1G 073403-5G
102529-978EA 380 USD
102529-978 102529-980
N-(2-Chloropyridin-4-yl)acetamide ≥95%
N-(2-Chloropyridin-4-yl)acetamide

Hazard: Irritant

Formula: C₇H₇ClN₂O
MDL Number: MFCD00234294
CAS Number: 13602-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR