(R)-2-Methylpyrrolidine hydrochloride ≥95%

Supplier: Matrix Scientific

073402-1G 073402-5G
102529-974EA 380 USD
102529-974 102529-976
(R)-2-Methylpyrrolidine hydrochloride ≥95%
(R)-2-Methylpyrrolidine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClN
MW: 121.6085 g/mol
MDL Number: MFCD06796610
CAS Number: 135324-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR