5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-3-cyano-1-(2,6-dichloro-4-trifluoromethylphenyl)pyrazole, 5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-1H-pyrazole-3-carbonitrile, 5-Amino-1-(2,6-dichloro-4-trifluoromethylphenyl)-3-cycano pyrazole

073398-1G 073398-5G
102529-950EA 15.38 USD
102529-950 102529-952
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile ≥95%
5-Amino-1-(2,6-dichloro-4-(trifluoromethyl)phenyl)-1H-pyrazole-3-carbonitrile

Hazard: Irritant

Formula: C₁₁H₅Cl₂F₃N₄
MW: 321.0900 g/mol
MDL Number: MFCD00227537
CAS Number: 120068-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR