3-(Bromomethyl)-5-chlorobenzo[b]thiophene ≥95%

Supplier: Matrix Scientific

073397-1G 073397-5G 073397-10G
102529-944EA 72.31 USD
102529-944 102529-946 102529-948
3-(Bromomethyl)-5-chlorobenzo[b]thiophene ≥95%
3-(Bromomethyl)-5-chlorobenzo[b]thiophene

Hazard: Irritant

Formula: C₉H₆BrClS
MW: 261.5700 g/mol
MDL Number: MFCD00173769
CAS Number: 1198-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR