2-Amino-6-chloropyrimidin-4-ol hydrate ≥95%

Supplier: Matrix Scientific

073395-1G 073395-5G 073395-10G
102529-936EA 320 USD
102529-936 102529-938 102529-940
2-Amino-6-chloropyrimidin-4-ol hydrate ≥95%
2-Amino-6-chloropyrimidin-4-ol hydrate

Hazard: Irritant

Formula: C₄H₆ClN₃O₂
MDL Number: MFCD00149407
CAS Number: 410530-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR