Methyl-6-aminopyrazine-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 6-aminopyrazine-2-carboxylate

073394-250MG 073394-1G 073394-5G
102529-930EA 423.69 USD
102529-930 102529-932 102529-934
Methyl-6-aminopyrazine-2-carboxylate ≥95%
Methyl-6-aminopyrazine-2-carboxylate

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.1400 g/mol
MDL Number: MFCD10697802
CAS Number: 118853-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR