2-Nitro-5-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methyl-2-nitropyridine, 6-Nitro-3-picoline

073389-1G 073389-5G
102529-912EA 40 USD
102529-912 102529-914
2-Nitro-5-picoline ≥95%
2-Nitro-5-picoline

Hazard: Irritant

Formula: C₆H₆N₂O₂
MW: 138.1200 g/mol
MDL Number: MFCD00234182
CAS Number: 1074-38-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR