Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Dimethyl-3-oxo-pyrrolidine-1-carboxylic acid ethyl ester, Ethyl 2,2-dimethyl-3-oxopyrrolidine-1-carboxylate

073388-250MG 073388-1G 073388-5G
102529-906EA 456.92 USD
102529-906 102529-908 102529-910
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate ≥95%
Ethyl-2,2-dimethyl-3-oxopyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₉H₁₅NO₃
MW: 185.2200 g/mol
MDL Number: MFCD09835297
CAS Number: 106556-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR