8-Bromo-2-chloroquinazolin-4-amine ≥95%
Supplier: Matrix Scientific
073386-250MG
073386-1G
102529-898EA
710.77
USD
102529-898
102529-900
8-Bromo-2-chloroquinazolin-4-amine ≥95%
8-Bromo-2-chloroquinazolin-4-amine
Hazard: Irritant
Formula:
C₈H₅BrClN₃ MW: 258.5024 g/mol |
MDL Number:
MFCD11858275 CAS Number: 956100-62-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...