2-Chloro-6-nitrobenzoic acid ≥95%

Supplier: Matrix Scientific

073385-100G 073385-25G
102529-896EA 235.38 USD
102529-896 102833-628
2-Chloro-6-nitrobenzoic acid ≥95%
2-Chloro-6-nitrobenzoic acid

Hazard: Irritant

Formula: C₇H₄ClNO₄
Melting Pt: 238-240°C
MDL Number: MFCD00100425
CAS Number: 5344-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR