2-Methyl-8-quinolinol ≥95%
Supplier: Matrix Scientific
Synonyms:
8-Hydroxy-2-methylquinoline, 8-Hydroxyquinaldine, 2-Methyloxine
073383-10G
073383-100G
102529-886EA
36.92
USD
102529-886
102529-888
2-Methyl-8-quinolinol ≥95%
2-Methyl-8-quinolinol
Hazard: Irritant
Formula:
C₁₀H₉NO MW: 159.19 g/mol |
MDL Number:
MFCD00006765 CAS Number: 826-81-3 UN: 3077 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...