(S)-3-(1-Aminoethyl)benzonitrile hydrochloride ≥95%
Supplier: Matrix Scientific
073382-250MG
073382-1G
073382-5G
102529-880EA
640
USD
102529-880
102529-882
102529-884
(S)-3-(1-Aminoethyl)benzonitrile hydrochloride ≥95%
(S)-3-(1-Aminoethyl)benzonitrile hydrochloride
Hazard: Irritant
Formula:
C₉H₁₁ClN₂ MW: 182.65 g/mol |
MDL Number:
MFCD11101269 CAS Number: 2741870-03-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...