3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one ≥95%

Supplier: Matrix Scientific

073380-1G 073380-5G 073380-10G
102529-868EA 284.62 USD
102529-868 102529-870 102529-872
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one ≥95%
3,5-Dichloro-2,6-dimethylpyridin-4(3H)-one

Hazard: Irritant

Formula: C₇H₇Cl₂NO
MDL Number: MFCD22123390
CAS Number: 2971-90-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR