N-Methyl-4-chloro-7-azaindole ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-1-methyl-1H-pyrrolo[2,3-b]pyridine
073379-250MG
073379-1G
073379-5G
102529-862EA
498.46
USD
102529-862
102529-864
102529-866
N-Methyl-4-chloro-7-azaindole ≥95%
N-Methyl-4-chloro-7-azaindole
Hazard: Irritant
Formula:
C₈H₇ClN₂ |
CAS Number:
74420-05-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...