N-Tosyl-4-chloro-3-iodo-7-azaindole ≥95%

Supplier: Matrix Scientific

073378-250MG 073378-1G 073378-5G
102529-856EA 404.62 USD
102529-856 102529-858 102529-860
N-Tosyl-4-chloro-3-iodo-7-azaindole ≥95%
N-Tosyl-4-chloro-3-iodo-7-azaindole

Hazard: Irritant

Formula: C₁₄H₁₀ClIN₂O₂S
MW: 432.6639 g/mol
MDL Number: MFCD12964070
CAS Number: 869335-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR