Ethyl-2-chloro-6-hydroxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-chloro-6-hydroxybenzoate

073376-250MG 073376-1G 073376-5G
102529-844EA 456.92 USD
102529-844 102529-846 102529-848
Ethyl-2-chloro-6-hydroxybenzoate ≥95%
Ethyl-2-chloro-6-hydroxybenzoate

Hazard: Irritant

Formula: C₉H₉ClO₃
MDL Number: MFCD09038111
CAS Number: 112270-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR