6-Amino-7-bromobenzothiazole ≥95%

Supplier: Matrix Scientific

Synonyms: 7-bromo-1,3-benzothiazol-6-amine

073375-250MG 073375-1G 073375-5G
102529-838EA 355.38 USD
102529-838 102529-840 102529-842
6-Amino-7-bromobenzothiazole ≥95%
6-Amino-7-bromobenzothiazole

Hazard: Irritant

Formula: C₇H₅BrN₂S
MW: 229.1000 g/mol
MDL Number: MFCD03426350
CAS Number: 769-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR