(S)-4-(4-Aminobenzyl)oxazolidin-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: (4S)-4-[(4-Aminophenyl)methyl]-2-oxazolidinone, (S)-4-(4-Aminobenzyl)-2(1H)-oxazolidinone

073374-250MG 073374-1G 073374-5G
102529-832EA 569.23 USD
102529-832 102529-834 102529-836
(S)-4-(4-Aminobenzyl)oxazolidin-2-one ≥95%
(S)-4-(4-Aminobenzyl)oxazolidin-2-one

Hazard: Irritant

Formula: C₁₀H₁₂N₂O₂
MW: 192.22 g/mol
MDL Number: MFCD03411476
CAS Number: 152305-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR