5-Bromothiophene-3-carboxylic acid ≥95%
Supplier: Matrix Scientific
073372-1G
073372-5G
102529-822EA
92.31
USD
102529-822
102529-824
5-Bromothiophene-3-carboxylic acid ≥95%
5-Bromothiophene-3-carboxylic acid
Hazard: Irritant
Formula:
C₅H₃BrO₂S |
MDL Number:
MFCD08695074 CAS Number: 100523-84-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...