5-Bromo-6-methylpyrimidine-2,4-diol ≥95%

Supplier: Matrix Scientific

073371-1G 073371-5G
102529-818EA 320 USD
102529-818 102529-820
5-Bromo-6-methylpyrimidine-2,4-diol ≥95%
5-Bromo-6-methylpyrimidine-2,4-diol

Hazard: Irritant

Formula: C₅H₅BrN₂O₂
MW: 205.0094 g/mol
MDL Number: MFCD12964069
CAS Number: 15018-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR