1-(2-Amino-5-bromophenyl)ethanone ≥95%

Supplier: Matrix Scientific

073370-250MG 073370-1G 073370-5G
102529-810EA 355.38 USD
102529-810 102529-812 102529-814
1-(2-Amino-5-bromophenyl)ethanone ≥95%
1-(2-Amino-5-bromophenyl)ethanone

Hazard: Irritant

Formula: C₈H₈BrNO
MDL Number: MFCD09834638
CAS Number: 29124-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR