Ethyl-5-(tert-butyl)isoxazole-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-(tert-butyl)isoxazole-3-carboxylate

073369-250MG 073369-1G 073369-5G
102529-804EA 152.31 USD
102529-804 102529-806 102529-808
Ethyl-5-(tert-butyl)isoxazole-3-carboxylate ≥95%
Ethyl-5-(tert-butyl)isoxazole-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MW: 197.231 g/mol
MDL Number: MFCD00085057
CAS Number: 91252-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR