Methyl-5-aminonicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 5-aminonicotinate, Methyl 5-aminopyridine-3-carboxylate

073368-1G 073368-5G
102529-798EA 23.08 USD
102529-798 102529-800
Methyl-5-aminonicotinate ≥95%
Methyl-5-aminonicotinate

Hazard: Irritant/Refrigerate

Formula: C₇H₈N₂O₂
MDL Number: MFCD04038688
CAS Number: 36052-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR