Benzo[b]thiophen-3-amine hydrochloride ≥95%

Supplier: Matrix Scientific

073367-1G 073367-5G
102529-794EA 640 USD
102529-794 102529-796
Benzo[b]thiophen-3-amine hydrochloride ≥95%
Benzo[b]thiophen-3-amine hydrochloride

Hazard: Irritant

Formula: C₈H₈ClNS
MW: 185.67 g/mol
MDL Number: MFCD18157696
CAS Number: 3394-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR