5-Bromoimidazo[1,2-a]pyrazine ≥95%

Supplier: Matrix Scientific

073365-250MG 073365-1G 073365-5G
102529-784EA 640 USD
102529-784 102529-786 102529-788
5-Bromoimidazo[1,2-a]pyrazine ≥95%
5-Bromoimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
MDL Number: MFCD08460076
CAS Number: 87597-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR