(5-Bromopyridin-2-yl)hydrazine ≥95%
Supplier: Matrix Scientific
Synonyms:
(5-Bromo-2-pyridyl)hydrazine, 5-Bromo-2-hydrazinopyridine
073364-1G
073364-5G
073364-10G
102529-778EA
15.38
USD
102529-778
102529-780
102529-782
(5-Bromopyridin-2-yl)hydrazine ≥95%
(5-Bromopyridin-2-yl)hydrazine
Hazard: Irritant
Formula:
C₅H₆BrN₃ MW: 188.0252 g/mol |
MDL Number:
MFCD00513424 CAS Number: 77992-44-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...