2-Amino-5-bromopyrimidin-4-ol hydrobromide ≥95%

Supplier: Matrix Scientific

073363-1G 073363-5G
102529-774EA 533.85 USD
102529-774 102529-776
2-Amino-5-bromopyrimidin-4-ol hydrobromide ≥95%
2-Amino-5-bromopyrimidin-4-ol hydrobromide

Hazard: Irritant

Formula: C₄H₅Br₂N₃O
MW: 270.9100 g/mol
MDL Number: MFCD12923111
CAS Number: 1215597-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR