2-Chloro-3-methylpyrazine ≥90%

Supplier: Matrix Scientific

073362-1G 073362-5G 073362-25G
102529-768EA 364.62 USD
102529-768 102529-770 102551-832
2-Chloro-3-methylpyrazine ≥90%
2-Chloro-3-methylpyrazine

Hazard: Irritant

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD00023279
CAS Number: 95-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR