2-Ethyl-3-hydroxy-6-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: Emoxypine, 2-Ethyl-6-methyl-3-pyridinol

073360-1G 073360-5G 073360-10G
102529-756EA 23.08 USD
102529-756 102529-758 102529-760
2-Ethyl-3-hydroxy-6-methylpyridine ≥95%
2-Ethyl-3-hydroxy-6-methylpyridine

Hazard: Irritant

Formula: C₈H₁₁NO
MW: 137.1800 g/mol
MDL Number: MFCD00462409
CAS Number: 2364-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR