Methyl-6-methylnicotinate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 6-methylnicotinate
073358-100G
073358-25G
102529-748EA
75.38
USD
102529-748
102833-626
Methyl-6-methylnicotinate ≥95%
Methyl-6-methylnicotinate
Hazard: Irritant/Air Sensitive/Stored under Argon
Formula:
C₈H₉NO₂ Melting Pt: 34…37 °C |
MDL Number:
MFCD00006340 CAS Number: 5470-70-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...