3-Hydroxy-2-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Nitropyridin-3-ol, 2-Nitro-3-pyridinol

073356-100G 073356-25G
102529-738EA 89.23 USD
102529-738 102663-950
3-Hydroxy-2-nitropyridine ≥95%
3-Hydroxy-2-nitropyridine

Hazard: Irritant

Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
MDL Number: MFCD00006258
CAS Number: 15128-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR