6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one ≥95%

Supplier: Matrix Scientific

073355-1G 073355-5G
102529-732EA 707.69 USD
102529-732 102529-734
6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one ≥95%
6-Thioxo-6,9-dihydro-1H-purin-2(3H)-one

Hazard: Irritant

Formula: C₅H₄N₄OS
MW: 168.1765 g/mol
MDL Number: MFCD00143509
CAS Number: 2002-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR