4-Fluoro-2-methoxyaniline ≥95%

Supplier: Matrix Scientific

073354-1G 073354-5G
102529-724EA 26.15 USD
102529-724 102529-726
4-Fluoro-2-methoxyaniline ≥95%
4-Fluoro-2-methoxyaniline

Hazard: Irritant-Harmful

Formula: C₇H₈FNO
MDL Number: MFCD00077536
CAS Number: 450-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR