5-Methoxy-2-nitrobenzonitrile ≥95%

Supplier: Matrix Scientific

073350-1G 073350-5G 073350-10G
102529-704EA 355.38 USD
102529-704 102529-706 102529-708
5-Methoxy-2-nitrobenzonitrile ≥95%
5-Methoxy-2-nitrobenzonitrile

Hazard: Irritant

Formula: C₈H₆N₂O₃
MW: 178.1500 g/mol
MDL Number: MFCD09056823
CAS Number: 38469-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR