2-Methylpyrimidin-5-ol ≥95%

Supplier: Matrix Scientific

073345-250MG 073345-1G 073345-5G
102529-674EA 355.38 USD
102529-674 102529-676 102529-678
2-Methylpyrimidin-5-ol ≥95%
2-Methylpyrimidin-5-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD09879706
CAS Number: 35231-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR